missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Procalcitonin Protein

Product Code. 18720789
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18720789 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18720789 Supplier R&D Systems™ Supplier No. 9607PN050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ Recombinant Human Procalcitonin Protein CF is encoded by a single CALCA gene, and can result in 2 alternatively spliced products: calcitonin and calcitonin-gene related peptide (CGRP).

The Recombinant Human Procalcitonin Protein is derived from E. coli.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01258-1
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 796
Molecular Weight (g/mol) 14 kDa
Name ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, CALC1, CALCA, Calcitonin 1, CGRP, CT, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Katacalcin, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, PCT, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, Preprocalcitonin, sarcolemmal calcium pump
Quantity 50 μg
Source E. coli-derived human Procalcitonin protein Pro27-Asn141,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.