missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pro-neuregulin-2, membrane-bound isoform Protein
A DNA sequence encoding the Homo sapiens (Human) Pro-neuregulin-2, membrane-bound isoform, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6436-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171864.1 | |
Pro-neuregulin-2, membrane-bound isoform Protein | |
Research Use Only | |
NRG2 | |
Recombinant Protein | |
E. coli | |
CYSPSLKSVQ DQAYKAPVVV EGKVQGLVPA GGSSSNSTRE PPASGRVALV KVLDKWPLRS GGLQREQVIS VGSCVPLERN QRYIFFLEPT EQPLVFKTAF APLDTNGKNL KKEVGKILCT DCATRPKLKK MKSQTGQVGE KQSLKCEAAA GNPQPSYRWF KDGKELNRSR DIRIKYGNGR KNSRLQFNKV KVEDAGEYVC EAENILGKDT VRGRLYVNSV STTLSSWSGH ARKCNETAKS YCVNGGVCYY IEGINQLSCK CPNGFFGQRC LEKLPLRLYM PDPKQKAEEL YQKR | |
Greater than 90% as determined by SDS-PAGE. |
9542 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |