missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKAB1 Protein
A cDNA sequence encoding the PRKAB1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13139-50ug
Additional Details : Weight : 0.01000kg
Specifications
PRKAB1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGNTSSE RAALERHGGH KTPRRDSSGG TKDGDRPKIL MDSPEDADLF HSEEIKAPEK EEFLAWQHDL EVNDKAPAQA RPTVFRWTGG GKEVYLSGSF NNWSKLPLTR SHNNFVAILD LPEGEHQYKF FVDGQWTHDP SEPIVTSQLG TVNNIIQVKK TDFEVFDALM VDSQKCSDVS ELSSSPPGPY HQEPYVCKPE ERFRAPPILP PHLLQVILNK DTGISCDPAL LPEPNHVMLN HLYALSIKDG VMVLSATHRY KKKYVTTLLY KPI | |
Greater than 85% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PRKAB1 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |