missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP3CA Protein
A cDNA sequence encoding the PPP3CA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13128-50ug
Additional Details : Weight : 0.01000kg
Specifications
PPP3CA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH TGSMSEPKAI DPKLSTTDRV VKAVPFPPSH RLTAKEVFDN DGKPRVDILK AHLMKEGRLE ESVALRIITE GASILRQEKN LLDIDAPVTV CGDIHGQFFD LMKLFEVGGS PANTRYLFLG DYVDRGYFSI ECVLYLWALK ILYPKTLFLL RGNHECRHLT EYFTFKQECK IKYSERVYDA CMDAFDCLPL AALMNQQFLC VHGGLSPEIN TLDDIRKLDR FKEPPAYGPM CDILWSDPLE DFGNEKTQEH FTHNTVRGCS YFYSYPAVCE FLQHNNLLSI LRAHEAQDAG YRMYRKSQTT GFPSLITIFS APNYLDVYNN KAAVLKYENN VMNIRQFNCS PHPYWLPNFM DVFTWSLPFV GEKVTEMLVN VLNICSDDEL GSEEDGFDGA TAAARKEVIR NKIRAIGKMA RVFSVLREES ESVLTLKGLT PTGMLPSGVL SGGKQTLQSA IKGFSPQHKI TSFEEAKGLD RINERMPPRR DAMPSDANLN SINKALTSET NGTDSNGSNS SNIQ | |
Greater than 85.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PPP3CA solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 5mM DTT, 1mM EDTA and 20% glycerol. |