missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PPP2R2B GST (N-Term) Protein

Product Code. 18302799 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18302799 10 μg 10µg
18341049 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18302799 Supplier Novus Biologicals™ Supplier No. H00005521Q0110ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 5521
Molecular Weight (g/mol) 36.63 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias B55BETA, FLJ95686, MGC24888, PP2A subunit B isoform B55-beta, PP2A subunit B isoform beta, PP2A subunit B isoform PR55-beta, PP2A subunit B isoform R2-beta, PP2A, subunit B, B-beta isoform, PP2AB55BETA, PP2ABBETA, PP2APR55B, PP2APR55BETA, PR2AB55BETA, PR2ABBETA, PR2APR55BETA, PR52B, PR55BETA, PR55-BETA, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, beta isoform, protein phosphatase 2, regulatory subunit B, beta, SCA12, serine/threonine protein phosphatase 2A, 55 kDa regulatory subunit B, betaisoform, serine/threonine protein phosphatase 2A, neuronal isoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B betaisoform, spinocerebellar ataxia 12
Gene Symbol PPP2R2B
Research Category Stem Cell Markers
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein PPP2R2B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.