missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP1R3B Protein
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Unit Size:
1mg
25µg
5µg
Specifications
Specifications
| Name | PPP1R3B Protein |
| Quantity | 25 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMMAVDIE YRYNCMAPSL RQERFAFKIS PKPSKPLRPC IQLSSKNEAS GMVAPAVQEK KVKKRVSFAD NQGLALTMVK VFSEFDDPLD MPFNITELLD NIVSLTTAES ESFVLDFSQP SADYLDFRNR LQADHVCLEN CVLKDKAIAG TVKVQNLAFE KTVKIRMTFD TWKSYTDFPC QYVKDTYAGS DRDTFSFDIS LPEKIQSYER MEFAVYYECN GQTYWDSNRG KNYRIIRAEL KSTQGMTKPH SGPDLGISFD QFGSPRCSYG LFPEWPSYLG YEKLGPYY |
| Buffer | The PPP1R3B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction