missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPCDC Protein

A cDNA sequence encoding the PPCDC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13115-1mg

 View more versions of this product

Product Code. 15962549

  • £2975.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

60490
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MEPKASCPAA APLMERKFHV LVGVTGSVAA LKLPLLVSKL LDIPGLEVAV VTTERAKHFY SPQDIPVTLY SDADEWEMWK SRSDPVLHID LRRWADLLLV APLDANTLGK VASGICDNLL TCVMRAWDRS KPLLFCPAMN TAMWEHPITA QQVDQLKAFG YVEIPCVAKK LVCGDEGLGA MAEVGTIVDK VKEVLFQHSG FQQS
Greater than 95% as determined by SDS-PAGE.
PPCDC Protein
Research Use Only
PPCDC
Human
His
The PPCDC protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer(pH 8.0), 1mM DTT, 0.1M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers