missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Potassium channel subfamily K member 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Potassium channel subfamily K member 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9230-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001017424.1 | |
Potassium channel subfamily K member 2 Protein | |
Research Use Only | |
KCNK2 | |
Recombinant Protein | |
Yeast | |
MLPSASRERP GYRAGVAAPD LLDPKSAAQN SKPRLSFSTK PTVLASRVES DTTINVMKWK TVSTIFLVVV LYLIIGATVF KALEQPHEIS QRTTIVIQKQ TFISQHSCVN STELDELIQQ IVAAINAGII PLGNTSNQIS HWD | |
Greater than 90% as determined by SDS-PAGE. |
3776 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |