missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Poly [ADP-ribose] polymerase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Poly [ADP-ribose] polymerase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6468-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001609.2 | |
Poly [ADP-ribose] polymerase 1 Protein | |
Research Use Only | |
PARP11 | |
Recombinant Protein | |
E. coli | |
AESSDKLYRV EYAKSGRASC KKCSESIPKD SLRMAIMVQS PMFDGKVPHW YHFSCFWKVG HSIRHPDVEV DGFSELRWDD QQKVKKTAEA GGVTGKGQDG IGSKAEKTLG DFAAEYAKSN RSTCKGCMEK IEKGQVRLSK KMVDPEKPQL GMIDRWYHPG CFVKNREELG FRPEYSASQL KGFSLLATED KEALKKQLPG VKSEGKRKGD EVDGVDEVAK KKSKKEKDKD SKLEKALKAQ NDLIWNIKDE LKKVCSTNDL KELLIFNKQQ VPSGESAILD RVADGMVFGA LLPCEECSGQ LVFKSDAYYC TGDVTAWTKC MVKTQTPNRK | |
Greater than 90% as determined by SDS-PAGE. |
142 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |