missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human POLR2I Protein

A cDNA sequence encoding the POLR2I was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13101-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942509

  • £155.00 / 10µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

POLR2I Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMEPDGTY EPGFVGIRFC QECNNMLYPK EDKENRILLY ACRNCDYQQE ADNSCIYVNK ITHEVDELTQ IIADVSQDPT LPRTEDHPCQ KCGHKEAVFF QSHSARAEDA MRLYYVCTAP HCGHRWTE
Greater than 85% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
POLR2I protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers