missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human POLR1C Protein

A cDNA sequence encoding the POLR1C was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13095-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15962489

  • £151.37 / 10µg
Estimated Shipment: 22-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

POLR1C Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
POLR1C protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAASQAV EEMRSRVVLG EFGVRNVHTT DFPGNYSGYD DAWDQDRFEK NFRVDVVHMD ENSLEFDMVG IDAAIANAFR RILLAEVPTM AVEKVLVYNN TSIVQDEILA HRLGLIPIHA DPRLFEYRNQ GDEEGTEIDT LQFRLQVRCT RNPHAAKDSS DPNELYVNHK VYTRHMTWIP LGNQADLFPE GTIRPVHDDI LIAQLRPGQE IDLLMHCVKG IGKDHAKFSP VATASYRLLP DITLLEPVEG EAAEELSRCF SPGVIEVQEV QGKKVARVAN PRLDTFSREI FRNEKLKKVV RLARVRDHYI FSVESTGVLP PDVLVSEAIK VLMGKCRRFL DELDAVQMD
Videos
SDS
Documents
Certificates
Special Offers

Special Offers