missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PNMT Protein
Click to view available options
:
20ug; Carrier Free
Description
The Recombinant Human PNMT Protein is derived from E. coli. The Recombinant Human PNMT Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, EDTA, Brij-35 and Glycerol. |
| Gene ID (Entrez) | 5409 |
| Molecular Weight (g/mol) | 32 kDa |
| Name | PNMT |
| Quantity | 20μg |
| Source | E. coli-derived human PNMT protein Met1-Leu282,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction