missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human PlGF Protein

Product Code. 18360288
Change view
Click to view available options
Quantity:
10 μg
50μg
Unit Size:
10µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360288 10 μg 10µg
18198250 50μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18360288 Supplier R&D Systems™ Supplier No. 264PGB010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Human VEGF R1/Flt-1 Fc Chimera (Catalog # 3516-FL ) is immobilized at 0.5 μg/mL, 100 μL/well, the concentration of Recombinant Human PlGF that produces 50% of the optimal binding response is approximately 0.75-4.5 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001193941.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 um filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Reconstitute (at 200 ug/mL in 4 mM HCl.)
Gene ID (Entrez) 5228
Molecular Weight (g/mol) Observed M.W.: 12-15 kDa, reducing conditions, Theoretical M.W.: 15 kDa
Quantity 10 μg
Source E. coli-derived human PlGF protein Ala21-Arg149
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 ug of the protein by the LAL method.
Gene Alias D12S1900, PGF, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760
Common Name PLGF
Gene Symbol PGF
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human VEGF R1/Flt-1 Fc Chimera is immobilized (at 0.5 ug/mL, 100 uL/well, the concentration of Recombinant Human PlGF that produces 50% of the optimal binding response is approximately 0.15-0.9 ng/mL)
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Ala21-Arg149
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.