missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PLGF His Protein

Product Code. 18395592 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.01 mg
0.02 mg
0.05 mg
0.1 mg
0.25 mg
Unit Size:
0.01mg
0.02mg
0.05mg
0.10mg
0.25mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395592 0.05 mg 0.05mg
18381852 0.01 mg 0.01mg
18304423 0.02 mg 0.02mg
18327072 0.1 mg 0.10mg
18374783 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18395592 Supplier Novus Biologicals™ Supplier No. NBP3118470.05MG

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Highly purified. Generating reliable and reproducible results.

TRUSTED_SUSTAINABILITY

Especificaciones

Concentration 0.25 mg/ml
For Use With (Application) Functional Assay
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 5228
Molecular Weight (g/mol) 18.3 kDa
Quantity 0.05 mg
Storage Requirements Store at -20°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration < 1 EU per 1ug of protein (determined by LAL method)
Gene Alias D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760
Gene Symbol PGF
Research Category Angiogenesis
Purity or Quality Grade >95%, by SDS-PAGE
Protein PLGF
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.