missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Pleiotrophin/PTN Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
Pleiotrophin/PTN protein is fully biologically active when compared to standard. The biological activity was measured by its ability to enhance neurite outgrowth of E16E18 rat embryonic cortical neurons, when neurons were plated on 96 well culture plates that had been precoated with 100 μL/well of a solution of 5-10 μg/mL rHuPTN.
Specifications
Specifications
| Accession Number | P21246, P21246, P21246 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 5764 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 15.3 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 0.1 EU/μg of Pleiotrophin/PTN as determined by LAL method. |
| Gene Alias | HARP, HBBM, HB-GAM, HBGF8, HBNF, HBNF1, HBNF-1, Heparin-binding brain mitogen, NEGF1HBGF-8, neurite growth-promoting factor 1, neurite growth-promoting factor1), OSF-1, Osteoblast-specific factor 1, pleiotrophin |
| Gene Symbol | PTN |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction