missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PLCD4 Protein

A DNA sequence encoding the Homo sapiens (Human) PLCD4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8084-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022571

  • £1735.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_116115.1
PLCD4 Protein
Research Use Only
PLCD4
Recombinant Protein
E. coli
MASLLQDQLT TDQDLLLMQE GMPMRKVRSK SWKKLRYFRL QNDGMTVWHA RQARGSAKPS FSISDVETIR NGHDSELLRS LAEELPLEQG FTIVFHGRRS NLDLMANSVE EAQIWMRGLQ LLVDLVTSMD HQERLDQWLS DWFQRGDKNQ DGKMSFQEVQ RLLHLMNVEM DQEYAFSLFQ AADTSQSGTL EGEEFVQFYK ALTKRAEVQE LFESFSADGQ KLTLLEFLDF LQEEQKERDC TSELALELID RYEPSDSGKL RHVLSMDGFL SYLCSKDGDI FNPACLPIYQ DMTQPLNHYF ICSSHNTYLV GDQLCGQSSV EGYIRALKRG CRCVEVDVWD GPSGEPVVYH GHTLTSRILF KDVVATVAQY AFQTSDYPVI LSLETHCSWE QQQTMARHLT EILGEQLLST TLDGVLPTQL PSPEELRRKI LVKGKKLTLE EDLEYEEEEA EPELEESELA LESQFETEPE PQEQNLQNKD KKKKSKPILC PALSSLVIYL KSVSFRSFTH SKEHYHFYEI SSFSETKAKR LIKEAGNEFV QHNTWQLSRV YPSGLRTDSS NYNPQELWNA GCQMVAMNMQ TAGLEMDICD GHFRQNGGCG YVLKPDFLRD IQSSFHPEKP ISPFKAQTLL IQVISGQQLP KVDKTKEGSI VDPLVKVQIF GVRLDTARQE TNYVENNGFN PYWGQTLCFR VLVPELAMLR FVVMDYDWKS RNDFIGQYTL PWTCMQQGYR HIHLLSKDGI SLRPASIFVY ICIQEGLEGD ES
Greater than 90% as determined by SDS-PAGE.
84812
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers