missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Placental Lactogen/CSH1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 5757-PL-025
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human Placental Lactogen/CSH1 Protein is derived from CHO. The Recombinant Human Placental Lactogen/CSH1 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
23.1 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
1442 | |
Placental Lactogen/CSH1 | |
Chinese Hamster Ovary cell line,CHO-derived human Placental Lactogen/CSH1 protein Val27-Phe217 with a C-terminal 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |