missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PKLR / PKRL Protein
A DNA sequence encoding the Homo sapiens (Human) PKLR / PKRL, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP1281-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000289.1 | |
PKLR / PKRL Protein | |
Research Use Only | |
Pklr | |
Recombinant Protein | |
E. coli | |
MSIQENISSL QLRSWVSKSQ RDLAKSILIG APGGPAGYLR RASVAQLTQE LGTAFFQQQQ LPAAMADTFL EHLCLLDIDS EPVAARSTSI IATIGPASRS VERLKEMIKA GMNIARLNFS HGSHEYHAES IANVREAVES FAGSPLSYRP VAIALDTKGP EIRTGILQGG PESEVELVKG SQVLVTVDPA FRTRGNANTV WVDYPNIVRV VPVGGRIYID DGLISLVVQK IGPEGLVTQV ENGGVLGSRK GVNLPGAQVD LPGLSEQDVR DLRFGVEHGV DIVFASFVRK ASDVAAVRAA LGPEGHGIKI ISKIENHEGV KRFDEILEVS DGIMVARGDL GIEIPAEKVF LAQKMMIGRC NLAGKPVVCA TQMLESMITK PRPTRAETSD VANAVLDGAD CIMLSGETAK GNFPVEAVKM QHAIAREAEA AVYHRQLFEE LRRAAPLSRD PTEVTAIGAV EAAFKCCAAA IIVLTTTGRS AQLLSRYRPR AAVIAVTRSA QAARQVHLCR GVFPLLYREP PEAIWADDVD RRVQFGIESG KLRGFLRVGD LVIVVTGWRP GSGYTNIMRV LSIS | |
Greater than 90% as determined by SDS-PAGE. |
5313 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |