missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PKC alpha Protein

Product Code. 18111830 Shop All Bio Techne Products
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 18111830

Brand: Novus Biologicals™ NBP1998835UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant protein for Human PKC alpha The Recombinant Human PKC alpha Protein is derived from Sf 9 (baculovirus). The Recombinant Human PKC alpha Protein has been validated for the following applications: SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.1 mg/mL
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol
Gene ID (Entrez) 5578
Molecular Weight (g/mol) 103 kDa
Quantity 5 μg
Source Sf 9 (baculovirus)
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha
Gene Symbol PRKCA
Buffer 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.