missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PISD Protein
A DNA sequence encoding the Homo sapiens (Human) PISD, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8162-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001313340.1 | |
PISD Protein | |
Research Use Only | |
PISD | |
Recombinant Protein | |
E. coli | |
MMCQSEARQG PELRAAKWLH FPQLALRRRL GQLSCMSRPA LKLRSWPLTV LYYLLPFGAL RPLSRVGWRP VSRVALYKSV PTRLLSRAWG RLNQVELPHW LRRPVYSLYI WTFGVNMKEA AVEDLHHYRN LSEFFRRKLK PQARPVCGLH SVISPSDGRI LNFGQVKNCE VEQVKGVTYS LESFLGPRMC TEDLPFPPAA SCDSFKNQLV TREGNELYHC VIYLAPGDYH CFHSPTDWTV SHRRHFPGSL MSVNPGMARW IKELFCHNER VVLTGDWKHG FFSLTAVGAT NVGSIRIYFD RDLHTNSPRH SKGSYNDFSF VTHTNREGVP MRKGEHLGEF NLGSTIVLIF EAPKDFNFQL KTGQKIRFGE ALGSL | |
Greater than 90% as determined by SDS-PAGE. |
23761 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |