missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PIH1D1 Protein

A cDNA sequence encoding the PIH1D1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13052-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942359

  • £155.00 / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PIH1D1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMANPKLL GMGLSEAEAI GADSARFEEL LLQASKELQQ AQTTRPESTQ IQPQPGFCIK TNSSEGKVFI NICHSPSIPP PADVTEEELL QMLEEDQAGF RIPMSLGEPH AELDAKGQGC TAYDVAVNSD FYRRMQNSDF LRELVITIAR EGLEDKYNLQ LNPEWRMMKN RPFMGSISQQ NIRSEQRPRI QELGDLYTPA PGRAESGPEK PHLNLWLEAP DLLLAEVDLP KLDGALGLSL EIGENRLVMG GPQQLYHLDA YIPLQINSHE SKAAFHRKRK QLMVAMPLLP VPS
Greater than 85% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PIH1D1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers