missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PHPT1 / PHP14 Protein

A DNA sequence encoding the Homo sapiens (Human) PHPT1 / PHP14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6491-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003680

  • £384.00 / 100µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001129333.1
PHPT1 / PHP14 Protein
Research Use Only
PHPT1
Recombinant Protein
E. coli
MAVADLALIP DVDIDSDGVF KYVLIRVHSA PRSGAPAAES KEIVRGYKWA EYHADIYDKV SGDMQKQGCD CECLGGGRIS HQSQDKKIHV YGYSMAYGPA QHAISTEKIK AKYPDYEVTW ANDGY
Greater than 90% as determined by SDS-PAGE.
29085
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers