missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PHOX2A GST (N-Term) Protein

Product Code. 18327567 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327567 10 μg 10µg
18314328 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18327567 Supplier Novus Biologicals™ Supplier No. H00000401Q0110ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Tekniske data

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 401
Molecular Weight (g/mol) M.W. (Theoretical): 35.64 kDa
Quantity 10 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias aristaless (Drosophila) homeobox, aristaless homeobox (Drosophila), fibrosis ofextraocular muscles, congenital, 2, autosomal recessive, aristaless homeobox homolog, Aristaless homeobox protein homolog, arix homeodomain protein, ARIX1 homeodomain protein, ARIXNCAM2, CFEOM2MGC52227, paired mesoderm homeobox protein 2A, paired-like homeobox 2apaired-like (aristaless) homeobox 2a, PMX2AFEOM2
Gene Symbol PHOX2A
Research Category Growth and Development, Neuronal Cell Markers, Neuroscience
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein PHOX2A
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.