missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PGEA1 GST (N-Term) Protein

Product Code. p-200052951 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 18318059

Brand: Novus Biologicals™ H00025776P0125ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 25776
Molecular Weight (g/mol) 40.9 kDa
Quantity 25 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias arb1, ARPP-binding protein, C22orf2chromosome 22 open reading frame 2, Cby, CBYgolgi and endoplasmic reticulum associated 1, chibby homolog 1 (Drosophila), coiled-coil protein PIGEA-14, Cytosolic leucine-rich protein, HS508I15A, PGEA1, PIGEA-14Chibby, PKD2 interactor, Golgi and endoplasmic reticulum-associated 1, polycystin-2 interactor, Golgi- and endoplasmic reticulum-associated protein, protein chibby homolog 1
Gene Symbol CBY1
Research Category Golgi Apparatus Markers
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein PGEA1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.