missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PFDN5 / MM-1 Protein
A DNA sequence encoding the Homo sapiens (Human) PFDN5 / MM-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8367-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002615.2 | |
PFDN5 / MM-1 Protein | |
Research Use Only | |
PFDN5 | |
Recombinant Protein | |
E. coli | |
AQSINITELN LPQLEMLKNQ LDQEVEFLST SIAQLKVVQT KYVEAKDCLN VLNKSNEGKE LLVPLTSSMY VPGKLHDVEH VLIDVGTGYY VEKTAEDAKD FFKRKIDFLT KQMEKIQPAL QEKHAMKQAV MEMMSQKIQQ LTALGAAQAT AKA | |
Greater than 90% as determined by SDS-PAGE. |
5204 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |