missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Persephin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 2388-PS-010/CF
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human Persephin Protein is derived from E. coli. The Recombinant Human Persephin Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | |
10.3 kDa (monomer) | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
5623 | |
Persephin | |
E. coli-derived human Persephin protein Ala61-Gly156 | |
Disulfide-linked homodimer | |
Recombinant |