missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Peroxiredoxin 1 / PRDX1 Protein

A DNA sequence encoding the Homo sapiens (Human) Peroxiredoxin 1 / PRDX1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8352-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16074171

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001189360.1
Peroxiredoxin 1 / PRDX1 Protein
Research Use Only
PRDX1
Recombinant Protein
E. coli
MSSGNAKIGH PAPNFKATAV MPDGQFKDIS LSDYKGKYVV FFFYPLDFTF VCPTEIIAFS DRAEEFKKLN CQVIGASVDS HFCHLAWVNT PKKQGGLGPM NIPLVSDPKR TIAQDYGVLK ADEGISFRGL FIIDDKGILR QITVNDLPVG RSVDETLRLV QAFQFTDKHG EVCPAGWKPG SDTIKPDVQK SKEYFSKQK
Greater than 90% as determined by SDS-PAGE.
5052
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers