missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals™ Recombinant Human Perilipin-2/ADFP Protein
Shop alle Bio Techne produkter
Klik for at se tilgængelige muligheder
Quantity:
0.5 mg
100 μg
Pakningsstørrelse:
0.5mg
100µg
Beskrivelse
A denatured recombinant protein with a N-Terminal T7-tag and corresponding to the amino acids 1-437 of Human Perilipin-2/ADFP The Recombinant Human Perilipin-2/ADFP Protein is derived from E. coli. The Recombinant Human Perilipin-2/ADFP Protein has been validated for the following applications: SDS-Page.
Tekniske data
Tekniske data
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 0.4M urea, 10% glycerol |
| Gene ID (Entrez) | 123 |
| Molecular Weight (g/mol) | 49.3kDa |
| Name | Human Perilipin-2/ADFP Protein |
| Purification Method | >85%, by SDS-PAGE |
| Quantity | 100 μg |
| Source | E.Coli |
| Immunogen | MASMTGGQQM GRGSMASVAV DPQPSVVTRV VNLPLVSSTY DLMSSAYLST KDQYPYLKSV CEMAENGVKT ITSVAMTSAL PIIQKLEPQI AVANTYACKG LDRIEERLPI LNQPSTQIVA NAKGAVTGAK DAVTTTVTGA KDSVASTITG VMDKTKGAVT GSVEKTKSVV SGSINTVLGS RMMQLVSSGV ENALTKSELL VEQYLPLTEE ELEKEAKKVE GFDLVQKPSY YVRLGSLSTK LHSRAYQQAL SRVKEAKQKS QQTISQLHST VHLIEFARKN VYSANQKIQD AQDKLYLSWV EWKRSIGYDD TDESHCAEHI ESRTLAIARN LTQQLQTTCH TLLSNIQGVP QNIQDQAKHM GVMAGDIYSV FRNAASFKEV SDSLLTSSKG QLQKMKESLD DVMDYLVDNT PLNWLVGPFY PQLTESQNAQ DQGAEMDKSS QETQRSEHKT H |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion