missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Perilipin-2/ADFP Protein

Product Code. 18207490 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100 μg
Unit Size:
0.5mg
100µg
This item is not returnable. View return policy

Product Code. 18207490

Brand: Novus Biologicals™ NBP2530390.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal T7-tag and corresponding to the amino acids 1-437 of Human Perilipin-2/ADFP The Recombinant Human Perilipin-2/ADFP Protein is derived from E. coli. The Recombinant Human Perilipin-2/ADFP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.4M urea, 10% glycerol
Gene ID (Entrez) 123
Molecular Weight (g/mol) 49.3kDa
Name Human Perilipin-2/ADFP Protein
Purification Method >85%, by SDS-PAGE
Quantity 0.5 mg
Source E.Coli
Immunogen MASMTGGQQM GRGSMASVAV DPQPSVVTRV VNLPLVSSTY DLMSSAYLST KDQYPYLKSV CEMAENGVKT ITSVAMTSAL PIIQKLEPQI AVANTYACKG LDRIEERLPI LNQPSTQIVA NAKGAVTGAK DAVTTTVTGA KDSVASTITG VMDKTKGAVT GSVEKTKSVV SGSINTVLGS RMMQLVSSGV ENALTKSELL VEQYLPLTEE ELEKEAKKVE GFDLVQKPSY YVRLGSLSTK LHSRAYQQAL SRVKEAKQKS QQTISQLHST VHLIEFARKN VYSANQKIQD AQDKLYLSWV EWKRSIGYDD TDESHCAEHI ESRTLAIARN LTQQLQTTCH TLLSNIQGVP QNIQDQAKHM GVMAGDIYSV FRNAASFKEV SDSLLTSSKG QLQKMKESLD DVMDYLVDNT PLNWLVGPFY PQLTESQNAQ DQGAEMDKSS QETQRSEHKT H
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2
Gene Symbol PLIN2
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.