missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Pentraxin 3/TSG-14 His-tag Protein

Product Code. 18758446
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18758446 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18758446 Supplier R&D Systems™ Supplier No. 10292TS050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Human C1q is immobilized at 5 μg/mL (100 μL/well), Recombinant Human Pentraxin 3/TSF-14 His-tag binds with an ED50 of 0.5 to 4 μg/mL
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P26022
For Use With (Application) Bioactivity
Gene ID (Entrez) 5806
Molecular Weight (g/mol) 41 kDa
Name alpha-induced protein 5, pentaxin-related gene, rapidly induced by IL-1 beta, tumor necrosis factor, Pentaxin-related protein PTX3, pentraxin 3, long, pentraxin-3, pentraxin-related gene, rapidly induced by IL-1 beta, pentraxin-related protein PTX3, PTX3, TNF alpha-induced protein 5, TNFAIP5, TSG14, TSG-14, TSG-14pentaxin-related gene, rapidly induced by IL-1 beta, Tumor necrosis factor alpha-induced protein 5, tumor necrosis factor, alpha-induced protein 5, Tumor necrosis factor-inducible gene 14 protein, tumor necrosis factor-inducible protein TSG-14
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Pentraxin 3/TSG-14 protein Glu18-Ser381, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.