missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDGFB Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 3 ng/mL, corresponding to a specific activity of >3.3 x 10^5 IU/mg.
Specifications
Specifications
| Accession Number | P01127, P01127, P01127 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 5155 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 24.8 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of PDGF-BB as determined by LAL method. |
| Gene Alias | c-sis, IBGC5, PDGF2, PDGF-2, PDGFB, Platelet-derived growth factor subunit B, SIS, SSV |
| Gene Symbol | PDGFB |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?