missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PDGF-BB Protein
Description
Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 1.5-6 ng/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 um filtered solution in Acetonitrile and TFA. |
| Gene ID (Entrez) | 5155 |
| Molecular Weight (g/mol) | 12.3 kDa (monomer) |
| Name | PDGF-BB |
| Quantity | 1 mg |
| Source | E. coli-derived human PDGF-BB protein Ser82-Thr190 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?