missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Recombinant Human PDGF-BB (CHO-expressed) Protein, CF

Product Code. 30058549
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 30058549

Brand: R&DSystems™ 11565PB050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 2.50-25.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01127.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 5155
Molecular Weight (g/mol) M.W.-observed: 11-13 kDa, M.W.-theroretical: 12 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human PDGF-BB protein Ser82-Thr190
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias c-sis;IBGC5;PDGF2;PDGF-2;PDGFB;Platelet-derived growth factor subunit B;SIS;SSV, PDGFBB
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in 4mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PDGF-BB
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.