missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDGF-AB Protein

Recombinant Protein

Brand:  enQuireBio™ QP5201-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945659

  • £2425.00 / 500µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PDGF-AB Protein
Research Use Only
PDGFA/B
Recombinant Protein
E. coli
Alphachain :MSIEEAVPA VCKTRTVIYE IPRSQVDPTS ANFLIWPPCV EVKRCTGCCN TSSVKCQPSR VHHRSVKVAK VEYVRKKPKL KEVQVRLEEH LECACATTSL NPDYREEDTG RPRESGKKRK RKRLKPT Bet achain:MSL GSLTIAEPAM IAECKTRTEV FEISRRLIDR TNANFLVWPP CVEVQRCSGC CNNRNVQCRP TQVQLRPVQV RKIEIVRKKP IFKKATVTLE DHLACKCETV AAARPVT
Greater than 95% as determined by SDS-PAGE.
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 20 ng/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^4 units/mg.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers