missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PDGF-AA Protein
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
The Recombinant Human PDGF-AA Protein is derived from E. coli. The Recombinant Human PDGF-AA Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
| Gene ID (Entrez) | 5154 |
| Molecular Weight (g/mol) | M.W. theroretical: 14 kDa (monomer) |
| Name | PDGF-AA |
| Quantity | 10 μg |
| Source | E. coli-derived human PDGF-AA protein Ser87-Thr211,with and without an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | PDGF1, PDGF-A, PDGFAA |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction