missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PD-L1/B7-H1 Fc Alexa Fluor™ 488 Protein
Click to view available options
:
20 μg
Description
Measured by flow cytometry for its ability to bind anti-human PD-L1/B7-H1 Monoclonal Antibody conjugated beads. The concentration of Recombinant Human PD-L1/B7-H1 Fc Chimera Alexa Fluor™ 488 (Catalog # AFG156) that produces 50% of the binding response is 15.0-150 ng/mL.
Specifications
Specifications
| Accession Number | Q9NZQ7.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS and NaCl with BSA as a carrier protein. |
| Gene ID (Entrez) | 29126 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 70-75 kDa, under reducing conditions., MolecularWeight-theroretical: 52 kDa (monomer) |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human PD-L1/B7-H1 protein Human PD-L1 (Phe19-Thr239) (N-terminus) Accession # Q9NZQ7.1 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction