missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human PD-1 Fc Chimera Protein

Product Code. 18390368
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390368 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390368 Supplier R&D Systems™ Supplier No. 1086PD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When Recombinant Human PD-1 Fc Chimera is immobilized at 0.1 μg/mL (100 μL/well), Recombinant Human B7-H1/PD-L1 Fc Chimera (Catalog # 156-B7 ) binds with a typical ED50 of 0.15-0.75 μg/mL.

Specifications

Accession Number Q15116.3
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5133
Molecular Weight (g/mol) M.W. (Observed): 60 - 70 kDa, under reducing conditions.
Name ASP/C3a desArg
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human PD-1 protein Human PD-1 (Leu25-Gln167) (C-terminus) Accession # Q15116.3 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Common Name ASP/C3a desArg
Gene Symbol PDCD1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.