missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PD-1 Fc Chimera Alexa Fluor™ 647 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by flow cytometry for its ability to bind anti-human PD-1 Monoclonal Antibody conjugated beads.The concentration of Recombinant Human PD-1 Fc Chimera Alexa Fluor™ 647 (Catalog # AFR1086) that produces 50% of the binding response is 0.25-5.00 ng/mL.
Specifications
Specifications
| Accession Number | Q15116.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 5133 |
| Molecular Weight (g/mol) | M.W.-observed: 60-70 kDa, under reducing conditions., M.W.-theroretical: 42.6 kDa (monomer) |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human PD-1 protein Human PD-1 (Leu25-Gln167) (N-terminus) Accession # Q15116.3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction