missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PCMTD1 Protein
A cDNA sequence encoding the PCMTD1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12977-2ug
Additional Details : Weight : 0.01000kg
Specifications
PCMTD1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGGAVSA GEDNDDLIDN LKEAQYIRTE RVEQAFRAID RGDYYLEGYR DNAYKDLAWK HGNIHLSAPC IYSEVMEALK LQPGLSFLNL GSGTGYLSTM VGLILGPFGI NHGIELHSDV VEYAKEKLES FIKNSDSFDK FEFCEPAFVV GNCLQIASDS HQYDRIYCGA GVQKDHENYM KILLKVGGIL VMPIEDQLTQ IMRTGQNTWE SKNILAVSFA PLV | |
Greater than 95% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PCMTD1 solution (0.25 mg/ml) contains 20mM Tris-HCl (pH 8.0), 20% glycerol, 0.15M NaCl and 1mM DTT. |