missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PAM Protein
Description
The Recombinant Human PAM Protein is derived from NS0. The Recombinant Human PAM Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 5066 |
| Molecular Weight (g/mol) | 90 kDa |
| Name | Peptidylglycine alpha-Amidating Monooxygenase/PAM |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human PAM protein Met1-Val817,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?