missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PAFAH2 His Protein

Product Code. 18108519 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
This item is not returnable. View return policy

Product Code. 18108519

Brand: Novus Biologicals™ NBP2520460.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-392 of Human PAFAH2 The Recombinant Human PAFAH2 Protein is derived from E. coli. The Recombinant Human PAFAH2 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH7.4) containing 20% glycerol, 1mM DTT
Gene ID (Entrez) 5051
Molecular Weight (g/mol) TMW: 46.4kDa
Quantity 0.1 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias EC 3.1.1, EC 3.1.1.47, FLJ26025, HSD-PLA2, platelet-activating factor acetylhydrolase 2 (40kD), platelet-activating factor acetylhydrolase 2, 40kDa, platelet-activating factor acetylhydrolase 2, cytoplasmic, SD-PLA2, Serine-dependent phospholipase A2
Common Name PAFAH2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.