missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PADI4 Protein
Description
The Recombinant Human PADI4 Protein is derived from E. coli. The Recombinant Human PADI4 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Sodium Acetate, NaCl, DTT, EDTA and Glycerol. |
| Gene ID (Entrez) | 23569 |
| Molecular Weight (g/mol) | 87 kDa |
| Name | PADI4 |
| Quantity | 10 μg |
| Source | E. coli-derived human PADI4 protein Ala2-Pro663,with an N-terminal 11 kDa fusion protein,a thrombin cleavage linker and a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?