missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human p53-regulated apoptosis-inducing protein 1

A DNA sequence encoding the Homo sapiens (Human) p53-regulated apoptosis-inducing protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8199-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003231

  • £384.00 / 100µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001182123.1
p53-regulated apoptosis-inducing protein 1
Research Use Only
TP53AIP1
Recombinant Protein
E. coli
MGSSSEVSFR SAQASCSGAR RQGLGRGDQN LSVMPPNGRA QTHTPGWVSP CSENRDGLLP ATAPGRLCSH RGADIPSFQT HQDPVTASGS SELHADCPQF RALDRAGN
Greater than 80% as determined by SDS-PAGE.
63970
100 μg
Not determined.
Full Length of BC069399
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers