missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human p38 beta/MAPK11 His Protein
Shop All Bio Techne Products
Click to view available options
:
10μg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-364 of Human p38 beta/MAPK11 The Recombinant Human p38 beta/MAPK11 Protein is derived from E. coli. The Recombinant Human p38 beta/MAPK11 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_002742 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 20% glycerol, 100mM NaCl |
| Gene ID (Entrez) | 5600 |
| Molecular Weight (g/mol) | 43.8kDa |
| Name | p38 beta/MAPK11 Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Purity or Quality Grade | >90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction