missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human p21WAF1 Protein
A DNA sequence encoding the Homo sapiens (Human) p21WAF1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5807-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000380.1 | |
p21WAF1 Protein | |
Research Use Only | |
CDKN1A | |
Recombinant Protein | |
E. coli | |
SEPAGDVRQN PCGSKACRRL FGPVDSEQLS RDCDALMAGC IQEARERWNF DFVTETPLEG DFAWERVRGL GLPKLYLPTG PRRGRDELGG GRRPGTSPAL LQGTAEEDHV DLSLSCTLVP RSGEQAEGSP GGPGDSQGRK RRQTSMTDFY HSKRRLIFSK RKP | |
Greater than 90% as determined by SDS-PAGE. |
1026 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |