missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Osteoprotegerin Protein is fully biologically active when compared to standard. The ED50 as determined by neutralizing the stimulation of U937 cells is less than 10 ng/mL, corresponding to a specific activity of >1.0 x 10^5 IU/mg in the presence of 10 ng/mL soluble rHuRANKL (sRANKL).
Specifications
Specifications
| Accession Number | O00300, O00300, O00300 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB,150 mM NaCl, pH 6.0. |
| Gene ID (Entrez) | 4982 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 19.7 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of Osteoprotegerin/TNFRSF11B as determined by LAL method. |
| Gene Alias | OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b |
| Gene Symbol | TNFRSF11B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?