missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Osteoprotegerin/TNFRSF11B Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
Osteoprotegerin Protein is fully biologically active when compared to standard. The ED50 as determined by neutralizing the stimulation of U937 cells is less than 10 ng/mL, corresponding to a specific activity of >1.0 x 10^5 IU/mg in the presence of 10 ng/mL soluble rHuRANKL (sRANKL).
Specifications
Specifications
| Accession Number | O00300, O00300, O00300 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB,150 mM NaCl, pH 6.0. |
| Gene ID (Entrez) | 4982 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 19.7 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of Osteoprotegerin/TNFRSF11B as determined by LAL method. |
| Gene Alias | OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b |
| Gene Symbol | TNFRSF11B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction