missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human OR2B6 GST (N-Term) Protein

Product Code. 18399949 Shop All Bio Techne Products
Change view
Click to view available options
:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18399949 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399949 Supplier Novus Biologicals™ Supplier No. H00026212P012ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 26212
Molecular Weight (g/mol) 61.8 kDa
Quantity 2 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias dJ408B20.2, hs6M1-32, MGC120190, MGC129828, Olfactory receptor 2B1, Olfactory receptor 2B5, olfactory receptor 2B6, Olfactory receptor 5-40, Olfactory receptor 6-31, Olfactory receptor OR6-4, olfactory receptor, family 2, subfamily B, member 1 pseudogene, olfactory receptor, family 2, subfamily B, member 5, olfactory receptor, family 2, subfamily B, member 6, olfactory receptor, family 2, subfamily B, member 6 pseudogene, OR2B1, OR2B1P, OR5-40OR2B5, OR5-41, OR6-31OR2B6P
Gene Symbol OR2B6
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein OR2B6
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.