missing translation for 'onlineSavingsMsg'
Få mere at vide

Novus Biologicals™ Recombinant Human OR12D3 GST (N-Term) Protein

Artikelnummer. 18427229 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
10 μg
25 μg
Pakningsstørrelse:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18427229 10 μg 10µg
18442219 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18427229 Leverandør Novus Biologicals™ Leverandørnr. H00081797P0110ug

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Tekniske data

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 81797
Molecular Weight (g/mol) MolecularWeight-theroretical: 62.2 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias hs6M1-27, MGC119267, MGC125888, olfactory receptor 12D3, Olfactory receptor OR6-27, olfactory receptor, family 12, subfamily D, member 3
Gene Symbol OR12D3
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein OR12D3
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.