missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human OCIL/CLEC2d Fc Chimera Protein, CF
Click to view available options
:
50 μg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human CD161 Fc Chimera Protein (Catalog # 7448-CD ) is immobilized at 10 μg/mL (100 μL/well), the concentration of Recombinant Human OCIL/CLEC2d Fc Chimera that produces 50% of the optimal binding response is approximately 1.5-9 μg/mL.
Specifications
Specifications
| Accession Number | Q9UHP7.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 29121 |
| Molecular Weight (g/mol) | M.W. (Observed): 50-60 kDa, under reducing conditions, M.W. (Theoretical): 42 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human OCIL/CLEC2d protein MD Human IgG1 (Pro100-Lys330) IEGR Human OCIL/CLEC2d (Ala58-Val191) (N-terminus) Accession # Q9UHP7.1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CLAXLLT-1, CLEC2D, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1, LLT1Osteoclast inhibitory lectin, OCILmember D |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction